Learn More
2-C-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-2,5-anhydro-3,4-O-(1-methylethylidene)-D-altrononitrile, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-C-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-2,5-anhydro-3,4-O-(1-methylethylidene)-D-altrononitrile
Specifications
Specifications
| Chemical Name or Material | 2-C-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-2,5-anhydro-3,4-O-(1-methylethylidene)-D-altrononitrile |
| CAS | 1191237-80-5 |
| Analyte or Component Names | 2-C-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-2,5-anhydro-3,4-O-(1-methylethylidene)-D-altrononitrile |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H17 N5 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-C-(4-Aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-2,5-anhydro-3,4-O-(1-methylethylidene)-D-altrononitrile |
| InChI Formula | InChI=1S/C15H17N5O4/c1-14(2)23-11-9(5-21)22-15(6-16,12(11)24-14)10-4-3-8-13(17)18-7-19-20(8)10/h3-4,7,9,11-12,21H,5H2,1-2H3,(H2,17,18,19)/t9-,11-,12-,15+/m1/s1 |
| SMILES | CC1(C)O[C@@H]2[C@@H](CO)O[C@](C#N)([C@@H]2O1)c3ccc4c(N)ncnn34 |
| IUPAC Name | (3aR,4R,6R,6aR)-4-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-6-(hydroxymethyl)-2,2-dimethyl-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxole-4-carbonitrile |
| Show More |