Learn More
2-Bromo-6-chloroaniline, Mikromol™
Product Code. 30459652
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459652 4 x 25 mg -
30051564 25 mg 25mg
30051266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30459652 Supplier Mikromol Supplier No. MM0006.2200254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Bromo-6-chloroaniline, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Bromo-6-chloroaniline
CAS 59772-49-5
Analyte or Component Names 2-Bromo-6-chloroaniline
Shipping Condition Room Temperature
Molecular Formula C6 H5 Br Cl N
Certifications/Compliance ISO 9001
Synonym 2-Bromo-6-chloroaniline, 2-Bromo-6-chlorobenzenamine
InChI Formula InChI=1S/C6H5BrClN/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2
Solution Type Neat
SMILES Nc1c(Cl)cccc1Br
IUPAC Name 2-bromo-6-chloroaniline
Molecular Weight (g/mol) 206.47
Quantity 4 x 25 mg
Formula Weight 204.9294 g/mol