missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-chloro-4-fluorophenol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Bromo-6-chloro-4-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Bromo-6-chloro-4-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 38°C to 40°C |
| Quantity | 25 g |
| Formula Weight | 225.45 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Bromo-6-chloro-4-fluorophenol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction