Learn More
2-Bromo-4-chloro-6-fluorophenol, 97%, Thermo Scientific™
Product Code. 11386849
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11386849
Brand: Thermo Scientific Alfa Aesar H26770.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 886499-88-3
Molecular Formula C6H3BrClFO
Molecular Weight (g/mol) 225.44
MDL Number MFCD04115955
InChI Key VGDYUSDMWUDGAB-UHFFFAOYSA-N
Synonym pubchem10171, 2-bromo-4-chloro-6-fluorophenol, 2-bromanyl-4-chloranyl-6-fluoranyl-phenol
PubChem CID 3838744
IUPAC Name 2-bromo-4-chloro-6-fluorophenol
SMILES OC1=C(Br)C=C(Cl)C=C1F
Specifications
Refractive Index 1.582
Quantity 25 g
Formula Weight 225.45
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-chloro-6-fluorophenol