missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(2-thienyl)thiazole, 97%, Thermo Scientific™

Product Code. 15444818
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444818 10 g 10g
15434818 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444818 Supplier Thermo Scientific Alfa Aesar Supplier No. H58575.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125409-90-6
Molecular Formula C7H4BrNS2
Molecular Weight (g/mol) 246.14
MDL Number MFCD11220135
InChI Key XRXGEQOSIYEEKD-UHFFFAOYSA-N
Synonym 2-bromo-4-2-thienyl thiazole, 2-bromo-4-thiophen-2-yl-1,3-thiazole
PubChem CID 38996210
IUPAC Name 2-bromo-4-thiophen-2-yl-1,3-thiazole
SMILES C1=CSC(=C1)C2=CSC(=N2)Br

Specifications

Quantity 10 g
Beilstein 21223492
Formula Weight 246.14
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(2-thienyl)thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.