accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1125409-90-6

SSNBr

CAS RN 1125409-90-6

IUPAC Name: 2-bromo-4-(thiophen-2-yl)-1,3-thiazole
Synonyms: 2-bromo-4-(thiophen-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 246.14
Molecular Formula: C7H4BrNS2
InChi Key: XRXGEQOSIYEEKD-UHFFFAOYSA-N
SMILES: BrC1=NC(=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results