missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2-chlorophenyl)ethan-1-one, Tech., Thermo Scientific™

Product Code. p-2697772
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10223522 1 g Amber Glass Bottle 1g
10712513 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10223522 Supplier Thermo Scientific Maybridge Supplier No. SPB06335DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5000-66-8
Molecular Formula C8H6BrClO
Molecular Weight (g/mol) 233.489
MDL Number MFCD00832993
InChI Key WZWWEVCLPKAQTA-UHFFFAOYSA-N
Synonym 2-bromo-2'-chloroacetophenone, 2-bromo-1-2-chlorophenyl ethanone, 2-bromo-1-2-chlorophenyl ethan-1-one, 2'-chlorophenacyl bromide, 2-chlorophenacyl bromide, 2'-chloro-2-bromoacetophenone, bromomethyl 2-chlorophenyl ketone, 2-bromo-1-2-chlorophenyl-1-ethanone, zlchem 501, 2-chlorophenacylbromide
PubChem CID 2735785
IUPAC Name 2-bromo-1-(2-chlorophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)Cl

Specifications

Density 1.6020g/mL
Packaging Amber Glass Bottle
Linear Formula BrCH2C(O)C6H4Cl
Quantity 1 g
Formula Weight 233.49
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-(2-chlorophenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.