accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClOBr

CAS RN 5000-66-8

IUPAC Name: 2-bromo-1-(2-chlorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(2-chlorophenyl)ethanone
Molecular Weight (g/mol): 233.49
Molecular Formula: C8H6BrClO
InChi Key: WZWWEVCLPKAQTA-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)CBr
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)