missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyphenylacetonitrile, 98%, Thermo Scientific™

Product Code. 11394106
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394106 500 mg 500mg
11304116 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394106 Supplier Thermo Scientific Alfa Aesar Supplier No. L19240.ME
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92552-22-2
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00016389
InChI Key WAQPXLOPRNLIGZ-UHFFFAOYSA-N
Synonym 2-benzyloxyphenylacetonitrile, 2-benzyloxy phenyl acetonitrile, 2-benzyloxyacetonitrile, benzeneacetonitrile, 2-phenylmethoxy, 2-2-benzyloxy phenyl acetonitrile, 2-benzyloxybenzyl cyanide, acmc-20b9y2, 2-benzyloxy benzyl cyanide
PubChem CID 561222
SMILES N#CCC1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 75°C to 78°C
Quantity 500 mg
Beilstein 2731076
IUPAC Name 2-(2-phenylmethoxyphenyl)acetonitrile
Formula Weight 223.28
Percent Purity 98%
Chemical Name or Material 2-Benzyloxyphenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.