accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 92552-22-2

NO

CAS RN 92552-22-2

IUPAC Name: 2-[2-(benzyloxy)phenyl]acetonitrile
Synonyms: 2-[2-(benzyloxy)phenyl]acetonitrile
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: WAQPXLOPRNLIGZ-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CC=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results