missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzimidazolylthio)acetic acid hydrazide, 97%, Thermo Scientific™

Product Code. p-7023538
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11433417

Brand: Thermo Scientific Alfa Aesar A17198.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30065-27-1
Molecular Formula C9H10N4OS
Molecular Weight (g/mol) 222.27
MDL Number MFCD00171027
InChI Key UIHKXOZJFOTOCE-UHFFFAOYSA-N
Synonym 2-benzimidazolylthio acetic acid hydrazide, 2-1h-benzo d imidazol-2-yl thio acetohydrazide, 2-1h-1,3-benzodiazol-2-ylsulfanyl acetohydrazide, 2-1h-benzimidazol-2-ylthio acetohydrazide, 2-1h-benzimidazol-2-ylsulfanyl acetohydrazide, 2-1h-1,3-benzimidazol-2-ylsulfanyl acetohydrazide, 1h-benzoimidazol-2-ylsulfanyl-acetic acid hydrazide, 2-benzimidazol-2-ylthioacetohydrazide, 2-1h-benzo d imidazol-2-ylthio acetohydrazide
PubChem CID 676606
SMILES NNC(=O)CSC1=NC2=CC=CC=C2N1

Specifications

Melting Point ∼160°C (decomposition)
Quantity 5 g
IUPAC Name 2-(1H-benzimidazol-2-ylsulfanyl)acetohydrazide
Formula Weight 222.27
Percent Purity 97%
Chemical Name or Material 2-(Benzimidazolylthio)acetic acid hydrazide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.