accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 30065-27-1

H2NNHOSNHN

CAS RN 30065-27-1

IUPAC Name: 2-(1H-1,3-benzodiazol-2-ylsulfanyl)acetohydrazide
Synonyms: 2-(1H-1,3-benzodiazol-2-ylsulfanyl)acetohydrazide
Molecular Weight (g/mol): 222.27
Molecular Formula: C9H10N4OS
InChi Key: UIHKXOZJFOTOCE-UHFFFAOYSA-N
SMILES: NNC(=O)CSC1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results