missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-hydroxypyrimidine, 95%

Product Code. 10517990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10517990 5 g Glass bottle 5g
10668550 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10517990 Supplier Thermo Scientific Acros Supplier No. 434330050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143489-45-6 | C4H5N3O | 111.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143489-45-6
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD08062411
InChI Key NVYMOVCPYONOSF-UHFFFAOYSA-N
Synonym 2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8
PubChem CID 17750388
IUPAC Name 2-aminopyrimidin-5-ol
SMILES NC1=NC=C(O)C=N1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 198.6°C to 199°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 111.1
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Amino-5-hydroxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-hydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.