accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2NOHN

CAS RN 143489-45-6

IUPAC Name: 2-aminopyrimidin-5-ol
Synonyms: 2-aminopyrimidin-5-ol
Molecular Weight (g/mol): 111.1
Molecular Formula: C4H5N3O
InChi Key: NVYMOVCPYONOSF-UHFFFAOYSA-N
SMILES: NC1=NC=C(O)C=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results