Learn More
2-Amino-4-cyanopyridine, 97%
Product Code. 11369825
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369825 1 g 1g
11379825 5 g 5g
11389825 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11369825 Supplier Thermo Scientific Alfa Aesar Supplier No. L19841.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 42182-27-4 | C6H5N3 | 119.127 g/mol
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42182-27-4
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD03791310
InChI Key GEEAYLFEIFJFGP-UHFFFAOYSA-N
Synonym 2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine
PubChem CID 7015524
IUPAC Name 2-aminopyridine-4-carbonitrile
SMILES C1=CN=C(C=C1C#N)N
Specifications
Melting Point 148°C to 152°C
Quantity 1 g
UN Number UN3439
Beilstein 386393
Solubility Information Slightly soluble in water.
Formula Weight 119.13
Percent Purity 97%
Chemical Name or Material 2-Amino-4-cyanopyridine