CAS RN 42182-27-4
CAS RN 42182-27-4
IUPAC Name:
2-aminopyridine-4-carbonitrile
Synonyms:
2-aminopyridine-4-carbonitrile
Molecular Weight (g/mol):
119.13
Molecular Formula:
C6H5N3
InChi Key:
GEEAYLFEIFJFGP-UHFFFAOYSA-N
SMILES:
NC1=CC(=CC=N1)C#N
1
–
1
of
1
results
1
2-Amino-4-cyanopyridine, 97%
CAS: 42182-27-4 Molecular Formula: C6H5N3 Molecular Weight (g/mol): 119.127 MDL Number: MFCD03791310 InChI Key: GEEAYLFEIFJFGP-UHFFFAOYSA-N Synonym: 2-amino-4-cyanopyridine,2-aminoisonicotinonitrile,2-amino-isonicotinonitrile,pubchem7664,4-cyanopyridin-2-amine,4-cyano-2-aminopyridine,acmc-1art6,2-amino-4-cyano-pyridine,ksc490s3f,4-cyano-2-pyridinamine PubChem CID: 7015524 IUPAC Name: 2-aminopyridine-4-carbonitrile SMILES: C1=CN=C(C=C1C#N)N