missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diacetoxynaphthalene, 98%, Thermo Scientific™

Product Code. 11467194
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11467194

Brand: Thermo Scientific Alfa Aesar B21588.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22472-26-0
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.25
MDL Number MFCD00036008
InChI Key VRMXWHJBWOVBEJ-UHFFFAOYSA-N
Synonym 2,7-diacetoxynaphthalene, naphthalene-2,7-diyl diacetate, 7-acetyloxy naphthalen-2-yl acetate, acmc-1cqps, 2,7-naphthalenediol diacetate, 7-acetyloxy-2-naphthyl acetate, 2,7-naphthalenediol,2,7-diacetate, 7-acetyloxynaphthalen-2-yl acetate, 7-acetyloxynaphthalen-2-yl ethanoate, acetic acid 7-acetyloxy-2-naphthalenyl ester
PubChem CID 790833
SMILES CC(=O)OC1=CC2=CC(OC(C)=O)=CC=C2C=C1

Specifications

Melting Point 129°C to 132°C
Quantity 5 g
IUPAC Name 7-(acetyloxy)naphthalen-2-yl acetate
Formula Weight 244.25
Percent Purity 98%
Chemical Name or Material 2,7-Diacetoxynaphthalene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.