accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 22472-26-0

H3COOOCH3O

CAS RN 22472-26-0

IUPAC Name: 7-(acetyloxy)naphthalen-2-yl acetate
Synonyms: 7-(acetyloxy)naphthalen-2-yl acetate
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: VRMXWHJBWOVBEJ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC2=CC(OC(C)=O)=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results