Learn More
2,6-Diiodophenol, Mikromol™
Product Code. 17980856
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980856 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17980856 Supplier Mikromol Supplier No. MM0694.180025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,6-Diiodophenol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6-Diiodophenol
CAS 28177-54-0
Analyte or Component Names 2,6-Diiodophenol
Shipping Condition Room Temperature
Molecular Formula C6 H4 I2 O
Certifications/Compliance ISO 9001
Synonym Phenol, 2,6-diiodo-, 2,6-Diiodophenol
InChI Formula InChI=1S/C6H4I2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
SMILES Oc1c(I)cccc1I
IUPAC Name 2,6-diiodophenol
Molecular Weight (g/mol) 345.9
Quantity 25 mg
Formula Weight 345.8352 g/mol