Learn More
2,6-Dichloropyridine-3-boronic acid, 95%
Product Code. 15483677
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15483677
Brand: Thermo Scientific Alfa Aesar H27406.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 148493-34-9 | C5H4BCl2NO2 | 191.802 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 148493-34-9
Molecular Formula C5H4BCl2NO2
Molecular Weight (g/mol) 191.802
MDL Number MFCD03094989
InChI Key XBBLBQZAVMHEER-UHFFFAOYSA-N
Synonym 2,6-dichloropyridine-3-boronic acid, 2,6-dichloropyridin-3-yl boronic acid, 2,6-dichloro-3-pyridineboronic acid, 3-borono-2,6-dichloropyridine, 2,6-dichloropyridin-3-yl-3-boronic acid, 2,6-dichloro-3-pyridinyl-boronic acid, boronic acid, 2,6-dichloro-3-pyridinyl, 2,6-dichloropyridine-3-boronicacid, pubchem5039, acmc-209d0p
PubChem CID 2762712
IUPAC Name (2,6-dichloropyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)Cl)Cl)(O)O
Specifications
Quantity 250 mg
Formula Weight 191.81
Percent Purity 95%
Chemical Name or Material 2,6-Dichloropyridine-3-boronic acid