accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 148493-34-9

HOBOHClNCl

CAS RN 148493-34-9

IUPAC Name: (2,6-dichloropyridin-3-yl)boronic acid
Synonyms: 2,6-dichloropyridin-3-ylboronic acid
Molecular Weight (g/mol): 191.8
Molecular Formula: C5H4BCl2NO2
InChi Key: XBBLBQZAVMHEER-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)N=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)