missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-hydroxybenzaldehyde, 98%

Product Code. 11422030
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422030 1 g 1g
11432030 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422030 Supplier Thermo Scientific Alfa Aesar Supplier No. H28395.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60964-09-2 | C7H4Cl2O2 | 191.01 g/mol

As starting reagents proposed to use 4-hydroxy-2,6-dimethylbenzaldehyde and 2,6-dichloro-4-hydroxybenzaldehyde easily available by straightforward syntheses from commercial products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As starting reagents proposed to use 4-hydroxy-2,6-dimethylbenzaldehyde and 2,6-dichloro-4-hydroxybenzaldehyde easily available by straightforward syntheses from commercial products.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60964-09-2
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD06797919
InChI Key WWFRBIPLCLSKNH-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-dichloro-4-hydroxy, 2,6-dichloro-4-hydroxy benzaldehyde, benzaldehyde,2,6-dichloro-4-hydroxy, 2,6-bis chloranyl-4-oxidanyl-benzaldehyde
PubChem CID 528382
IUPAC Name 2,6-dichloro-4-hydroxybenzaldehyde
SMILES OC1=CC(Cl)=C(C=O)C(Cl)=C1

Specifications

Melting Point 224°C to 229°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-4-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.