accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60964-09-2

ClOClOH

CAS RN 60964-09-2

IUPAC Name: 2,6-dichloro-4-hydroxybenzaldehyde
Synonyms: 2,6-dichloro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: WWFRBIPLCLSKNH-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(C=O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results