Learn More
2-(5-Nitro-1H-imidazol-1-yl)ethanol, LoGiCal
Product Code. 30461335
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461335 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461335 Supplier LoGiCal Supplier No. LGCFOR0012.07
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(5-Nitro-1H-imidazol-1-yl)ethanol
CAS 5006-68-8
Analyte or Component Names 2-(5-Nitro-1H-imidazol-1-yl)ethanol
Shipping Condition Room Temperature
Molecular Formula C5 H7 N3 O3
Certifications/Compliance ISO 9001
Synonym 2-(5-Nitro-1H-imidazol-1-yl)ethanol, Metronidazole Imp. D (EP), 5-Nitro-1H-imidazole-1-ethanol, 5-Nitroimidazole-1-ethanol, 1-(2-Hydroxyethyl)-5-nitroimidazole, Metronidazole Impurity D
InChI Formula InChI=1S/C5H7N3O3/c9-2-1-7-4-6-3-5(7)8(10)11/h3-4,9H,1-2H2
Solution Type Neat
SMILES OCCn1cncc1[N+](=O)[O-]
IUPAC Name 2-(5-nitroimidazol-1-yl)ethanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 157.13
Quantity 10 mg
Formula Weight 157.0487 g/mol