Learn More
2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(benzyloxycarbonylamino)hexanoate (N,N'-Bis(benzyloxycarbonyl)-L-lysine N-Succinimidyl Ester), Mikromol™
Description
2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(benzyloxycarbonylamino)hexanoate (N,N'-Bis(benzyloxycarbonyl)-L-lysine N-Succinimidyl Ester)
Specifications
Specifications
| Chemical Name or Material | 2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(benzyloxycarbonylamino)hexanoate |
| CAS | 21160-83-8 |
| Analyte or Component Names | 2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(benzyloxycarbonylamino)hexanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H29 N3 O8 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Lysine, N2,N6-bis[(phenylmethoxy)carbonyl]-, 2,5-dioxo-1-pyrrolidinyl ester, Carbamic acid, [(1S)-1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(phenylmethyl) ester (9CI), Carbamic acid, [1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(phenylmethyl) ester, (S)-, Succinimide, N-[(N2,N6-dicarboxy-L-lysyl)oxy]-, dibenzyl ester (8CI), NSC 250409, Nalpha,Nepsilon-Dibenzyloxycarbonyl-L-lysine N-hydroxysuccinimide ester |
| InChI Formula | InChI=1S/C26H29N3O8/c30-22-14-15-23(31)29(22)37-24(32)21(28-26(34)36-18-20-11-5-2-6-12-20)13-7-8-16-27-25(33)35-17-19-9-3-1-4-10-19/h1-6,9-12,21H,7-8,13-18H2,(H,27,33)(H,28,34)/t21-/m0/s1 |
| SMILES | O=C(NCCCC[C@H](NC(=O)OCc1ccccc1)C(=O)ON2C(=O)CCC2=O)OCc3ccccc3 |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis(phenylmethoxycarbonylamino)hexanoate |
| Show More |