Learn More
2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-carboxylic Acid, Mikromol™
Description
2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 1-(3-Carboxy-2-pyridyl)-4-methyl-2-phenylpiperazine |
| CAS | 61338-13-4 |
| Analyte or Component Names | 1-(3-Carboxy-2-pyridyl)-4-methyl-2-phenylpiperazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(4-Methyl-2-phenyl-1-piperazinyl)-3-pyridinecarboxylic acid, 1-(3-Carboxy-2-pyridyl)-4-methyl-2-phenylpiperazine |
| InChI Formula | InChI=1S/C17H19N3O2/c1-19-10-11-20(15(12-19)13-6-3-2-4-7-13)16-14(17(21)22)8-5-9-18-16/h2-9,15H,10-12H2,1H3,(H,21,22) |
| SMILES | CN1CCN(C(C1)c2ccccc2)c3ncccc3C(=O)O |
| IUPAC Name | 2-(4-methyl-2-phenylpiperazin-1-yl)pyridine-3-carboxylic acid |
| Show More |