missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzyl chloride, 96%, Thermo Scientific™

Product Code. 11397845
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397845 5 g 5g
11307855 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397845 Supplier Thermo Scientific Alfa Aesar Supplier No. L02810.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-55-5
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
MDL Number MFCD00015556
InChI Key BETNPSBTDMBHCZ-UHFFFAOYSA-N
Synonym 2,4-dimethylbenzyl chloride, 1-chloromethyl-2,4-dimethylbenzene, 4-chloromethyl-m-xylene, benzene, 1-chloromethyl-2,4-dimethyl, 2,4-dimethyl-1-chloromethyl benzene, 1-chloromethyl-2,4-dimethyl-benzene, acmc-209tf3, timtec-bb sbb005790, labotest-bb ltbb004402
PubChem CID 69989
IUPAC Name 1-(chloromethyl)-2,4-dimethylbenzene
SMILES CC1=CC=C(CCl)C(C)=C1

Specifications

Density 1.06
Boiling Point 103°C (15 mmHg)
Flash Point 96°C (204°F)
Odor Pungent
Refractive Index 1.5386
Quantity 5 g
UN Number UN3265
Beilstein 742366
Formula Weight 154.64
Percent Purity 96%
Chemical Name or Material 2,4-Dimethylbenzyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.