accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClH3C

CAS RN 824-55-5

IUPAC Name: 1-(chloromethyl)-2,4-dimethylbenzene
Synonyms: 1-(chloromethyl)-2,4-dimethylbenzene
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: BETNPSBTDMBHCZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCl)C(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results