Learn More
2,4-Dimethyl-3-heptanone, 99%
Product Code. 11454297
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454297 10 g 10g
11464297 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11454297 Supplier Thermo Scientific Alfa Aesar Supplier No. B20384.09
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
CAS: 18641-71-9 | C9H18O | 142.242 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18641-71-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00027146
InChI Key MMEXTUAHJZOQKN-UHFFFAOYSA-N
PubChem CID 29178
IUPAC Name 2,4-dimethylheptan-3-one
SMILES CCCC(C)C(=O)C(C)C
Specifications
Density 0.812
Boiling Point 170°C
Flash Point 44°C (111°F)
Odor Pleasant
Quantity 10 g
UN Number UN1224
Synonym 2,4-dimethyl-3-heptanone, 3-heptanone, 2,4-dimethyl, 3-heptanone,2,4-dimethyl, acmc-1ca3p
Formula Weight 142.24
Percent Purity 99%
Chemical Name or Material 2,4-Dimethyl-3-heptanone