Learn More
2,4-Dihydroxy-7-methoxy-1,4-benzoxazine-3-one (DIMBOA) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30315391
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315391 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30315391 Supplier Dr. Ehrenstorfer Supplier No. DREA12634820AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DIMBOA
CAS 15893-52-4
Analyte or Component Names DIMBOA
Shipping Condition Room Temperature
Molecular Formula C9 H9 N O5
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3(4H)-one, (±)-2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3(4H)-one, (±)-DIMBOA, 2,3-Dihydro-2,4-dihydroxy-7-methoxy-4H-1,4-benzoxazin-3-one, 2,4-Dihydroxy-3-keto-7-methoxy-1,4-benzoxazine, 2,4-Dihydroxy-7-methoxy-1,4-(2H)-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-1,4-benzoxazine-3-one, 2,4-Dihydroxy-7-methoxy-2H-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-3,4-dihydro-2H-1,4-benzoxazin-3-one, 2,4-Dihydroxy-7-methoxy-4H-1,4-benzoxazin-3(2H)-one, DIMBOA, 2H-1,4-Benzoxazin-3(4H)-one, 2,4-dihydroxy-7-methoxy-
InChI Formula InChI=1S/C9H9NO5/c1-14-5-2-3-6-7(4-5)15-9(12)8(11)10(6)13/h2-4,9,12-13H,1H3
SMILES COc1ccc2N(O)C(=O)C(O)Oc2c1
IUPAC Name 2,4-dihydroxy-7-methoxy-1,4-benzoxazin-3-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 211.17
Quantity 1 mL
Formula Weight 211.0481 g/mol