missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis

Product Code. 11309136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11309136 5 g 5g
11399126 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11309136

Brand: Thermo Scientific Alfa Aesar L20266.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79784-36-4 | C8H4F2O2 | 170.12 g/mol

Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79784-36-4
Molecular Formula C8H4F2O2
Molecular Weight (g/mol) 170.12
MDL Number MFCD04038287
InChI Key AZBQYELTMQNVST-UHFFFAOYSA-N
Synonym 2,4-difluorophenylglyoxal hydrate, 2-2,4-difluorophenyl-2-oxoacetaldehyde hydrate, 2,4-difluorophenylglyoxal monohydrate, benzeneacetaldehyde,2,4-difluoro-a-oxo, 2-2,4-difluorophenyl-2-oxoethanal, hydrate, 2-2,4-bis fluoranyl phenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 2782300
IUPAC Name 2-(2,4-difluorophenyl)-2-oxoacetaldehyde;hydrate
SMILES FC1=CC(F)=C(C=C1)C(=O)C=O

Specifications

Melting Point 51°C to 54°C
Quantity 5 g
Beilstein 4387465
Solubility Information Soluble in water.
Formula Weight 170.12 (Anhydrous)
Percent Purity 95%
Assay (dry wt.)
Chemical Name or Material 2,4-Difluorophenylglyoxal hydrate

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.