accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 79784-36-4

IUPAC Name: 2-(2,4-difluorophenyl)-2-oxoacetaldehyde hydrate
Synonyms: 2-(2,4-difluorophenyl)-2-oxoacetaldehyde hydrate
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: BILNRTMGRXGXSV-UHFFFAOYSA-N
SMILES: O.FC1=CC(F)=C(C=C1)C(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results