Learn More
2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol Fumarate, Mikromol™
Description
2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol Fumarate
Specifications
Specifications
| Chemical Name or Material | 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol Fumarate |
| CAS | 2734403-85-9 |
| Analyte or Component Names | 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol Fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 N3 O S . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Quetiapine Fumarate Imp. I (EP), Quetiapine Imp. I (EP), 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol Fumarate, Quetiapine Fumarate Imp. I (EP) as Fumarate, Quetiapine Fumarate Impurity I as Fumarate, Quetiapine Impurity I as Fumarate |
| InChI Formula | InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H6 |
| SMILES | OCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24.OC(=O)\C=C\C(=O)O |
| IUPAC Name | 2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethanol;(E)-but-2-enedioic acid |
| Show More |