Learn More
2,4-D-iso-octyl ester (technical) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4-D-iso-octyl ester (technical) |
| CAS | 25168-26-7 |
| Analyte or Component Names | 2,4-D-iso-octyl ester (technical) |
| Shipping Condition | +20°C |
| Molecular Formula | C16 H22 Cl2 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Acetic acid, 2-(2,4-dichlorophenoxy)-, isooctyl ester, Acetic acid, (2,4-dichlorophenoxy)-, isooctyl ester (6CI,8CI,9CI), Isooctyl alcohol, (2,4-dichlorophenoxy)acetate (8CI), 2,4-D ester, 2,4-D isooctyl ester, 2,4-Dichlorophenoxyacetic acid isooctyl ester, Agro-D-Ester, Agroxone 5, Esteron, Isooctyl 2,4-dichlorophenoxyacetate, Silvapron D |
| Solvent or Matrix | Acetonitrile |
| Molecular Weight (g/mol) | 333.25 |
| Show More |