missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Biphenylyl)-2-propanol, 95%

Product Code. 11334584
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334584 50 g 50g
11324584 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334584 Supplier Thermo Scientific Alfa Aesar Supplier No. L01364.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34352-74-4 | C15H16O | 212.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34352-74-4
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00014390
InChI Key GOKGIYHIVSGXDM-UHFFFAOYSA-N
Synonym 2-4-biphenylyl-2-propanol, 2-4-phenylphenyl propan-2-ol, 2-biphenyl-4-yl propan-2-ol, 2-1,1'-biphenyl-4-yl propan-2-ol, 2-4-biphenyl-2-propanol, 1,1'-biphenyl-4-methanol, .alpha.,.alpha.-dimethyl, acmc-1aevv, cambridge id 5135301, 1,1'-biphenyl-4-methanol, alpha,alpha-dimethyl
PubChem CID 118654
IUPAC Name 2-(4-phenylphenyl)propan-2-ol
SMILES CC(C)(C1=CC=C(C=C1)C2=CC=CC=C2)O

Specifications

Melting Point 90°C to 93°C
Quantity 50 g
Beilstein 1947575
Formula Weight 212.29
Percent Purity 97%
Chemical Name or Material 2-(4-Biphenylyl)-2-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.