accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3COH

CAS RN 34352-74-4

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}propan-2-ol
Synonyms: 2-{[1,1'-biphenyl]-4-yl}propan-2-ol
Molecular Weight (g/mol): 212.29
Molecular Formula: C15H16O
InChi Key: GOKGIYHIVSGXDM-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results