missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromo-3-hydroxybenzoic acid, 97%

Product Code. 11430248
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430248 1 g 1g
11440248 5 g 5g
11450248 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430248 Supplier Thermo Scientific Alfa Aesar Supplier No. B23790.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14348-40-4 | C7H3Br3O3 | 374.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14348-40-4
Molecular Formula C7H3Br3O3
Molecular Weight (g/mol) 374.81
MDL Number MFCD00055557
InChI Key YDBHVMTTYXWHLI-UHFFFAOYSA-N
Synonym 3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid
PubChem CID 151915
IUPAC Name 2,4,6-tribromo-3-hydroxybenzoic acid
SMILES C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br

Specifications

Melting Point 144°C to 149°C
Quantity 1 g
Beilstein 2844497
Formula Weight 374.81
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromo-3-hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.