Learn More
2,4,5-Tribromotoluene, 97+%
Product Code. 11445603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445603 1 g 1g
11455603 5 g 5g
11465603 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11445603 Supplier Thermo Scientific Alfa Aesar Supplier No. A13507.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 3278-88-4 | C7H5Br3 | 328.829 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3278-88-4
Molecular Formula C7H5Br3
Molecular Weight (g/mol) 328.829
MDL Number MFCD00052984
InChI Key KZZJNNUPNNBCCH-UHFFFAOYSA-N
Synonym 2,4,5-tribromotoluene, benzene, 1,2,4-tribromo-5-methyl, acmc-209upl, 2,4,5-tribrom-toluol, ksc569e9t, benzene,1,2,4-tribromo-5-methyl, 1,2,4-tris bromanyl-5-methyl-benzene
PubChem CID 137875
IUPAC Name 1,2,4-tribromo-5-methylbenzene
SMILES CC1=CC(=C(C=C1Br)Br)Br
Specifications
Melting Point 112°C to 115°C
Odor Odorless
Quantity 1 g
Formula Weight 328.83
Percent Purity ≥97%
Chemical Name or Material 2,4,5-Tribromotoluene