accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3278-88-4

CH3BrBrBr

CAS RN 3278-88-4

IUPAC Name: 1,2,4-tribromo-5-methylbenzene
Synonyms: 1,2,4-tribromo-5-methylbenzene
Molecular Weight (g/mol): 328.83
Molecular Formula: C7H5Br3
InChi Key: KZZJNNUPNNBCCH-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(Br)C=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)