Learn More
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
Product Code. 15420497
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Product Code. 15420497
Brand: Thermo Scientific Alfa Aesar J66024.09
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 14227-66-8 | C26H35BrO17 | 699.45 g/mol
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Soluble in water. Slightly soluble in ethanol
Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14227-66-8
Molecular Formula C26H35BrO17
Molecular Weight (g/mol) 699.45
MDL Number MFCD00069839
InChI Key NLFHLQWXGDPOME-GLMDSCOTSA-N
Synonym cel br heptaacetate, alpha-d-cellobiosyl bromide heptaacetate, cellobiosyl bromide heptaacetate, acetobromo-d-cellobiose, 2,3,6,2',3',4',6'-hepta-o-acetyl-a-d-cellobiosyl bromide, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-2r,3r,4s,5r,6r-4,5-diacetoxy-2-acetoxymethyl-6-bromotetrahydro-2h-pyran-3-yl oxy tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, alpha-d-glucopyranosyl bromide, 4-o-2,3,4,6-tetra-o-acetyl-beta-d-glucopyranosyl-, triacetate
PubChem CID 11039776
SMILES CC(=O)OC[C@H]1OC(Br)C(OC(C)=O)[C@@H](OC(C)=O)C1O[C@@H]1OC(COC(C)=O)[C@@H](OC(C)=O)C(OC(C)=O)[C@H]1OC(C)=O
Specifications
Melting Point 180°C to 190°C
Color White
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water; Slightly soluble in ethanol
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate
Formula Weight 699.45
Physical Form Powder
Chemical Name or Material 2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide