accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14227-66-8

CH3OO(R)OBrOCH3O(S)OH3COO(S)OOCH3O(R)OCH3OOCH3O(R)OH3CO

CAS RN 14227-66-8

IUPAC Name: [(2R,4S)-4,5-bis(acetyloxy)-6-bromo-3-{[(2S,3R,5R)-3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Synonyms: [(2R,4S)-4,5-bis(acetyloxy)-6-bromo-3-{[(2S,3R,5R)-3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 699.45
Molecular Formula: C26H35BrO17
InChi Key: NLFHLQWXGDPOME-GLMDSCOTSA-N
SMILES: CC(=O)OC[C@H]1OC(Br)C(OC(C)=O)[C@@H](OC(C)=O)C1O[C@@H]1OC(COC(C)=O)[C@@H](OC(C)=O)C(OC(C)=O)[C@H]1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)