Learn More
2,3,5-Trichloroaniline 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30312619
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312619 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30312619 Supplier Dr. Ehrenstorfer Supplier No. DREA17700050AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3,5-Trichloroaniline
CAS 18487-39-3
Analyte or Component Names 2,3,5-Trichloroaniline
Shipping Condition +20°C
Molecular Formula C6H4NCl3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2,3,5-Trichloroaniline, Aniline, 2,3,5-trichloro- (6CI,8CI), 2,3,5-Trichlorobenzenamine
InChI Formula InChI=1S/C6H4Cl3N/c7-3-1-4(8)6(9)5(10)2-3/h1-2H,10H2
SMILES Nc1cc(Cl)cc(Cl)c1Cl
IUPAC Name 2,3,5-trichloroaniline
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 196.46
Quantity 1 mL
Formula Weight 194.9409 g/mol