Learn More
2,3,4,5-Tetrafluorobenzoic acid, 98+%, Thermo Scientific™
Product Code. 11433198
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433198 25 g 25g
11413198 1 g 1g
11423198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11433198 Supplier Thermo Scientific Alfa Aesar Supplier No. A18954.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1201-31-6
Molecular Formula C7H2F4O2
Molecular Weight (g/mol) 194.09
MDL Number MFCD00009613
InChI Key SFKRXQKJTIYUAG-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro benzoic acid, benzoic acid, tetrafluoro, pubchem1350, tetrafluorobenzoic acid, acmc-1bre4, ksc174o7d, sfkrxqkjtiyuag-uhfffaoysa, 2,3,4,5-tetrafluorbenzoic acid, 2,3,4,5-tetrafluoro-benzoic acid, 2,3,4,5-?tetrafluorobenzoic acid
PubChem CID 297549
IUPAC Name 2,3,4,5-tetrafluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(F)C(F)=C1F
Specifications
Melting Point 85°C to 87°C
Quantity 25 g
Beilstein 2052670
Formula Weight 194.09
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoic acid