CAS RN 1201-31-6
CAS RN 1201-31-6
IUPAC Name:
2,3,4,5-tetrafluorobenzoic acid
Synonyms:
2,3,4,5-tetrafluorobenzoic acid
Molecular Weight (g/mol):
194.09
Molecular Formula:
C7H2F4O2
InChi Key:
SFKRXQKJTIYUAG-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(F)=C(F)C(F)=C1F
1
–
2
of
2
results
2
2,3,4,5-Tetrafluorobenzoic acid, 98+%, Thermo Scientific™
CAS: 1201-31-6 Molecular Formula: C7H2F4O2 Molecular Weight (g/mol): 194.09 MDL Number: MFCD00009613 InChI Key: SFKRXQKJTIYUAG-UHFFFAOYSA-N Synonym: 2,3,4,5-tetrafluoro benzoic acid,benzoic acid, tetrafluoro,pubchem1350,tetrafluorobenzoic acid,acmc-1bre4,ksc174o7d,sfkrxqkjtiyuag-uhfffaoysa,2,3,4,5-tetrafluorbenzoic acid,2,3,4,5-tetrafluoro-benzoic acid,2,3,4,5-?tetrafluorobenzoic acid PubChem CID: 297549 IUPAC Name: 2,3,4,5-tetrafluorobenzoic acid SMILES: OC(=O)C1=CC(F)=C(F)C(F)=C1F