Learn More
2,2'-[Oxybis[butane-1,4-diyl-(piperazine-1,4-diyl)]]dipyrimidine, Mikromol™
Description
2,2'-[Oxybis[butane-1,4-diyl-(piperazine-1,4-diyl)]]dipyrimidine
Specifications
Specifications
| Chemical Name or Material | 2,2'-[Oxybis[butane-1,4-diyl-(piperazine-1,4-diyl)]]dipyrimidine |
| CAS | 2724726-67-2 |
| Analyte or Component Names | 2,2'-[Oxybis[butane-1,4-diyl-(piperazine-1,4-diyl)]]dipyrimidine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H38 N8 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Buspirone Hydrochloride Imp. D (EP), Buspirone Imp. D (EP), 2,2'-[Oxybis[butane-1,4-diyl-(piperazine-1,4-diyl)]]dipyrimidine, Buspirone Hydrochloride Impurity D, Buspirone Impurity D |
| InChI Formula | InChI=1S/C23H32ClN3.2ClH/c1-25(2)14-6-15-26(3)16-7-17-27-22-9-5-4-8-19(22)10-11-20-12-13-21(24)18-23(20)27;;/h4-5,8-9,12-13,18H,6-7,10-11,14-17H2,1-3H3;2*1H |
| SMILES | C(CCN1CCN(CC1)c2ncccn2)COCCCCN3CCN(CC3)c4ncccn4 |
| IUPAC Name | 2-[4-[4-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butoxy]butyl]piperazin-1-yl]pyrimidine |
| Show More |