missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenyl)thioacetamide, 97%, Thermo Scientific™

Product Code. 15484137
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15484137 5 g 5g
15474137 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15484137

Brand: Thermo Scientific Alfa Aesar H31541.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1053656-06-6
Molecular Formula C9H11NOS
Molecular Weight (g/mol) 181.253
MDL Number MFCD09757577
InChI Key DIECFUUYDGQVBU-UHFFFAOYSA-N
Synonym 2-2-methoxyphenyl thioacetamide, 2-2-methoxyphenyl ethanethioamide, 1-amino-2-2-methoxyphenyl ethane-1-thione
PubChem CID 14718040
IUPAC Name 2-(2-methoxyphenyl)ethanethioamide
SMILES COC1=CC=CC=C1CC(=S)N

Specifications

Melting Point 86°C to 89°C
Quantity 5 g
Formula Weight 181.27
Percent Purity 97%
Chemical Name or Material 2-(2-Methoxyphenyl)thioacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.