accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1053656-06-6

CH3OSNH2

CAS RN 1053656-06-6

IUPAC Name: 2-(2-methoxyphenyl)ethanethioamide
Synonyms: 2-(2-methoxyphenyl)ethanethioamide
Molecular Weight (g/mol): 181.25
Molecular Formula: C9H11NOS
InChi Key: DIECFUUYDGQVBU-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1CC(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results