Learn More
2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one, Mikromol™
Description
2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one
Spezifikation
Spezifikation
| Chemical Name or Material | 2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one |
| CAS | 1218-69-5 |
| Analyte or Component Names | 2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H9 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4H-1,3-Benzoxazin-4-one, 2-(2-hydroxyphenyl)-, 4H-1,3-Benzoxazin-4-one, 2-(o-hydroxyphenyl)- (7CI,8CI), 2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one, 2-(2-Hydroxyphenyl)benz[e][1,3]oxazin-4-one, 2-(2'-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one, 2-(2-Hydroxyphenyl)-4H-1,3-benzoxazin-4-one |
| InChI Formula | InChI=1S/C14H9NO3/c16-11-7-3-1-5-9(11)14-15-13(17)10-6-2-4-8-12(10)18-14/h1-8,16H |
| SMILES | Oc1ccccc1C2=NC(=O)c3ccccc3O2 |
| IUPAC Name | 2-(2-hydroxyphenyl)-1,3-benzoxazin-4-one |
| Mehr anzeigen |