Learn More
2,2-Dimethylbutanoic Acid-d6, TRC
Product Code. 30384089
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384089 250 mg 250mg
30387454 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30384089 Supplier Toronto Research Chemicals Supplier No. TRCD468042250MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101419-75-4
Molecular Formula C6H6D6O2
Molecular Weight (g/mol) 122.2
Synonym 2,2-Dimethylbutyric Acid-d6, Neohexanoic Acid-d6, α,α-Dimethylbutanoic Acid-d6, α,α-Dimethylbutyric Acid-d6, NSC 16045-d6, NSC 741804-d6
IUPAC Name 2,2-bis(methyl-d3)butanoic acid
SMILES OC(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])CC)=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C6H12O2/c1-4-6(2,3)5(7)8/h4H2,1-3H3,(H,7,8)/i2D3,3D3
Formula Weight 122.12
Chemical Name or Material 2,2-Dimethylbutanoic Acid-d6