CAS RN 101419-75-4
CAS RN 101419-75-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-Dimethylbutanoic Acid-d6, TRC
CAS: 101419-75-4 Molecular Formula: C6H6D6O2 Molecular Weight (g/mol): 122.2 Synonym: 2,2-Dimethylbutyric Acid-d6,Neohexanoic Acid-d6,α,α-Dimethylbutanoic Acid-d6,α,α-Dimethylbutyric Acid-d6,NSC 16045-d6,NSC 741804-d6 IUPAC Name: 2,2-bis(methyl-d3)butanoic acid SMILES: OC(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])CC)=O